Naringin hydrate, 98%
Thermo Scientific Chemicals

Naringin hydrate, 98%

CAS: 132203-74-8 | C27H32O14 | 580.54 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number L10163.09
also known as L10163-09
Price (USD)
27.60
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
27.60
Each
Chemical Identifiers
CAS132203-74-8
SpecificationsSpecification SheetSpecification Sheet
Formpowder
Water Content (Karl Fischer Titration)≤ 5.0%
Optical Rotation-90? ? 3? (c=1, ethanol)
Appearance (Color)white to cream or pale yellow
Assay (unspecified)> 97.5%
A citrus bioflavanoid that inhibits cytochrome P450 monooxygenase activity

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only