Isopropylmalonic acid, 98+%
Isopropylmalonic acid, 98+%
Isopropylmalonic acid, 98+%
Thermo Scientific Chemicals

Isopropylmalonic acid, 98+%

CAS: 601-79-6 | C6H10O4 | 146.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L10171.06
also known as L10171-06
Price (USD)
67.65
Special offer
Online exclusive
Ends: 28-Dec-2026
79.50
Save 11.85
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
67.65
Special offer
Online exclusive
Ends: 28-Dec-2026
79.50
Save 11.85
Each
Chemical Identifiers
CAS601-79-6
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Melting Point (clear melt)86.5-92.5?C
Assay (Aqueous acid-base Titration)≥98.0 to ≤102.0%
Appearance (Color)White to pale cream
Assay (Silylated GC)≥98.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only