4-Cyanophenyl isothiocyanate, 98%
4-Cyanophenyl isothiocyanate, 98%
4-Cyanophenyl isothiocyanate, 98%
Thermo Scientific Chemicals

4-Cyanophenyl isothiocyanate, 98%

CAS: 2719-32-6 | C8H4N2S | 160.194 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L10173.03
also known as L10173-03
Price (USD)
27.65
Special offer
Online exclusive
Ends: 30-Jun-2026
32.10
Save 4.45 (14%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
27.65
Special offer
Online exclusive
Ends: 30-Jun-2026
32.10
Save 4.45 (14%)
Each
Chemical Identifiers
CAS2719-32-6
IUPAC Name4-isothiocyanatobenzonitrile
Molecular FormulaC8H4N2S
InChI KeyDZFKAXLNKZXNHD-UHFFFAOYSA-N
SMILESS=C=NC1=CC=C(C=C1)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)117-123?C
Appearance (Color)Cream or pale yellow to yellow-brown
Assay (GC)≥97.5%
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only