2,6-Dichlorobenzothiazole, 97%
2,6-Dichlorobenzothiazole, 97%
2,6-Dichlorobenzothiazole, 97%
Thermo Scientific Chemicals

2,6-Dichlorobenzothiazole, 97%

CAS: 3622-23-9 | C7H3Cl2NS | 204.068 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L10256.03
also known as L10256-03
Price (USD)
28.10
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
28.10
Each
Chemical Identifiers
CAS3622-23-9
IUPAC Name2,6-dichloro-1,3-benzothiazole
Molecular FormulaC7H3Cl2NS
InChI KeyQDZGJGWDGLHVNK-UHFFFAOYSA-N
SMILESClC1=NC2=CC=C(Cl)C=C2S1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
Assay (GC)≥96.0%
Melting Point (clear melt)92.0-98.0?C
FormCrystals or powder or crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only