Tetrahydrofurfuryl acrylate, tech. 90%, stab.
Tetrahydrofurfuryl acrylate, tech. 90%, stab.
Tetrahydrofurfuryl acrylate, tech. 90%, stab.
Thermo Scientific Chemicals

Tetrahydrofurfuryl acrylate, tech. 90%, stab.

CAS: 2399-48-6 | C8H12O3 | 156.181 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number L10299.18
also known as L10299-18
Price (USD)
33.65
Special offer
Online exclusive
Ends: 28-Dec-2026
39.30
Save 5.65
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
33.65
Special offer
Online exclusive
Ends: 28-Dec-2026
39.30
Save 5.65
Each
Chemical Identifiers
CAS2399-48-6
IUPAC Name(oxolan-2-yl)methyl prop-2-enoate
Molecular FormulaC8H12O3
InChI KeyYNXCGLKMOXLBOD-UHFFFAOYNA-N
SMILESC=CC(=O)OCC1CCCO1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Assay (GC)≥88.0%
Refractive Index1.4500-1.4600 @ 20?C
Stabilizerca. 500ppm hydroquinone (HQ) or ca. 500pm 4-methoxyphenol/methylhydroquinone (MEHQ)
Identification (FTIR)Conforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only