2-Chloro-6-methylthiophenol, 96%
2-Chloro-6-methylthiophenol, 96%
2-Chloro-6-methylthiophenol, 96%
2-Chloro-6-methylthiophenol, 96%
Thermo Scientific Chemicals

2-Chloro-6-methylthiophenol, 96%

CAS: 18858-05-4 | C7H7ClS | 158.643 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number L10499.03
also known as L10499-03
Price (USD)
46.65
Online Exclusive
51.80
Save 5.15 (10%)
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
46.65
Online Exclusive
51.80
Save 5.15 (10%)
Each
Add to cart
Chemical Identifiers
CAS18858-05-4
IUPAC Name2-chloro-6-methylbenzene-1-thiol
Molecular FormulaC7H7ClS
InChI KeyWNRLJMYSWRBJIG-UHFFFAOYSA-N
SMILESCC1=C(S)C(Cl)=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥95.0%
Appearance (Color)Clear colorless to yellow
Refractive Index1.5920-1.5980 @ 20?C
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only