1,3-Cyclooctadiene, 97%
1,3-Cyclooctadiene, 97%
1,3-Cyclooctadiene, 97%
Thermo Scientific Chemicals

1,3-Cyclooctadiene, 97%

CAS: 1700-10-3 | C8H12 | 108.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L10516.14
also known as L10516-14
Price (USD)
124.00
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
124.00
Each
Chemical Identifiers
CAS1700-10-3
IUPAC Name(1Z,3Z)-cycloocta-1,3-diene
Molecular FormulaC8H12
InChI KeyRRKODOZNUZCUBN-CCAGOZQPSA-N
SMILESC1CC\C=C/C=C\C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥96.0%
Identification (FTIR)Conforms
Refractive Index1.4910-1.4960 @ 20°C
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only