Ethyl pentafluorobenzoate, 98%
Ethyl pentafluorobenzoate, 98%
Ethyl pentafluorobenzoate, 98%
Ethyl pentafluorobenzoate, 98%
Thermo Scientific Chemicals

Ethyl pentafluorobenzoate, 98%

CAS: 4522-93-4 | C9H5F5O2 | 240.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L10523.14
also known as L10523-14
Price (USD)
89.65
Special offer
Online exclusive
Ends: 30-Jun-2026
105.00
Save 15.35 (15%)
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
89.65
Special offer
Online exclusive
Ends: 30-Jun-2026
105.00
Save 15.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS4522-93-4
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
FormLiquid
Appearance (Color)Clear colorless to yellow
Refractive Index1.4270-1.4310 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only