Ethyl pipecolinate, 98+%
Ethyl pipecolinate, 98+%
Ethyl pipecolinate, 98+%
Thermo Scientific Chemicals

Ethyl pipecolinate, 98+%

CAS: 15862-72-3 | C8H16NO2 | 158.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number L10532.03
also known as L10532-03
Price (USD)
29.70
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
29.70
Each
Chemical Identifiers
CAS15862-72-3
IUPAC Name(2S)-2-(ethoxycarbonyl)piperidin-1-ium
Molecular FormulaC8H16NO2
InChI KeySZIKRGHFZTYTIT-ZETCQYMHSA-O
SMILESCCOC(=O)[C@@H]1CCCC[NH2+]1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Refractive Index1.4540-1.4580 @ 20?C
Appearance (Color)Clear colorless to yellow
Assay (GC)≥98.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only