3-(4-Methylbenzoyl)acrylic acid, 98%
3-(4-Methylbenzoyl)acrylic acid, 98%
3-(4-Methylbenzoyl)acrylic acid, 98%
Thermo Scientific Chemicals

3-(4-Methylbenzoyl)acrylic acid, 98%

CAS: 20972-36-5 | C11H10O3 | 190.198 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L10551.03
also known as L10551-03
Price (USD)
26.65
Online Exclusive
29.80
Save 3.15 (11%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
26.65
Online Exclusive
29.80
Save 3.15 (11%)
Each
Chemical Identifiers
CAS20972-36-5
IUPAC Name4-(4-methylphenyl)-4-oxobut-2-enoic acid
Molecular FormulaC11H10O3
InChI KeyVNJMEFZKYZHWEO-UHFFFAOYSA-N
SMILESCC1=CC=C(C=C1)C(=O)C=CC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale yellow to yellow
FormCrystals or powder or crystalline powder
Assay (HPLC)≥97.5%
Melting Point (clear melt)136.0-142.0?C
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only