4,4'-Dimethoxychalcone, 99%
4,4'-Dimethoxychalcone, 99%
4,4'-Dimethoxychalcone, 99%
4,4'-Dimethoxychalcone, 99%
Thermo Scientific Chemicals

4,4'-Dimethoxychalcone, 99%

CAS: 2373-89-9 | C17H16O3 | 268.31 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number L10585.18
also known as L10585-18
Price (USD)
160.65
Special offer
Online exclusive
Ends: 30-Jun-2026
189.00
Save 28.35 (15%)
Each
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)
160.65
Special offer
Online exclusive
Ends: 30-Jun-2026
189.00
Save 28.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS2373-89-9
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale yellow to yellow
Assay (GC)≥98.5%
Melting Point (clear melt)98-104?C
FormCrystals or powder or crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only