Diethylene glycol diacrylate, tech. 75%, stab.
Diethylene glycol diacrylate, tech. 75%, stab.
Diethylene glycol diacrylate, tech. 75%, stab.
Thermo Scientific Chemicals

Diethylene glycol diacrylate, tech. 75%, stab.

CAS: 4074-88-8 | C10H14O5 | 214.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number L10682.22
also known as L10682-22
Price (USD)
57.65
Online Exclusive
64.20
Save 6.55 (10%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
57.65
Online Exclusive
64.20
Save 6.55 (10%)
Each
Chemical Identifiers
CAS4074-88-8
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Assay from Suppliers CofA≥73.0%
Appearance (Color)Clear, colorless
Diethylene glycol diacrylate is used as reactive diluent and cross linking agent of radiation curing system, and also may be used as crosslinking resins, plastics and rubber modifier.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethylene glycol diacrylate is used as reactive diluent and cross linking agent of radiation curing system, and also may be used as crosslinking resins, plastics and rubber modifier.

Solubility
Slightly soluble in water (1-5g/100ml at 18°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. It is sensitive to light. Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Gregory M. Cruise.; David S. Scharp.; Jeffrey A. Hubbell. Characterization of permeability and network structure of interfacially photopolymerized poly(ethylene glycol) diacrylate hydrogels. Biomaterials. 1998, 19 (14), 1287-1294.
  2. Cruise GM.; Hegre OD.; Lamberti FV.; Hager SR.; Hill R.; Scharp DS.; Hubbell JA. In vitro and in vivo performance of porcine islets encapsulated in interfacially photopolymerized poly(ethylene glycol) diacrylate membranes. Cell Transplantation. 1999, 8 (3), 293-306.