2-Chloro-4-(trifluoromethyl)pyridine, 98+%
2-Chloro-4-(trifluoromethyl)pyridine, 98+%
2-Chloro-4-(trifluoromethyl)pyridine, 98+%
2-Chloro-4-(trifluoromethyl)pyridine, 98+%
Thermo Scientific Chemicals

2-Chloro-4-(trifluoromethyl)pyridine, 98+%

CAS: 81565-18-6 | C6H3ClF3N | 181.54 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L10713.03
also known as L10713-03
Price (USD)
71.65
Online Exclusive
79.10
Save 7.45 (9%)
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
71.65
Online Exclusive
79.10
Save 7.45 (9%)
Each
Add to cart
Chemical Identifiers
CAS81565-18-6
IUPAC Name2-chloro-4-(trifluoromethyl)pyridine
Molecular FormulaC6H3ClF3N
InChI KeyGBNPVXZNWBWNEN-UHFFFAOYSA-N
SMILESFC(F)(F)C1=CC=NC(Cl)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Refractive Index1.4470-1.4510 @ 20?C
FormLiquid
Assay (GC)≥98.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only