Hexafluoroacetone trihydrate, 98%
Hexafluoroacetone trihydrate, 98%
Hexafluoroacetone trihydrate, 98%
Thermo Scientific Chemicals

Hexafluoroacetone trihydrate, 98%

CAS: 34202-69-2 | C3H6F6O4 | 220.067 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number L10777.22
also known as L10777-22
Price (USD)
324.65
Online Exclusive
361.00
Save 36.35 (10%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
324.65
Online Exclusive
361.00
Save 36.35 (10%)
Each
Chemical Identifiers
CAS34202-69-2
IUPAC Namehexafluoropropan-2-one trihydrate
Molecular FormulaC3H6F6O4
InChI KeySNZAEUWCEHDROX-UHFFFAOYSA-N
SMILESO.O.O.FC(F)(F)C(=O)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.3160-1.3220 @ 20°C
FormLiquid
Water Content (Karl Fischer Titration)21.0-30.0% (2.4-3.4 waters of hydration)
Appearance (Color)Clear colorless
Assay (GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only