1-Pyrenecarboxaldehyde, 98+%
1-Pyrenecarboxaldehyde, 98+%
1-Pyrenecarboxaldehyde, 98+%
Thermo Scientific Chemicals

1-Pyrenecarboxaldehyde, 98+%

CAS: 3029-19-4 | C17H10O | 230.266 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L10835.14
Price (USD)
195.65
Special offer
Online exclusive
Ends: 30-Jun-2026
230.00
Save 34.35 (15%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
195.65
Special offer
Online exclusive
Ends: 30-Jun-2026
230.00
Save 34.35 (15%)
Each
Chemical Identifiers
CAS3029-19-4
IUPAC Namepyrene-1-carbaldehyde
Molecular FormulaC17H10O
InChI KeyRCYFOPUXRMOLQM-UHFFFAOYSA-N
SMILESO=CC1=CC=C2C=CC3=CC=CC4=CC=C1C2=C34
View more
SpecificationsSpecification SheetSpecification Sheet
Free acid (titration)≤1%
Melting Point (clear melt)123.0-130.0?C
Appearance (Color)Dark yellow to yellow
FormCrystals or powder or crystalline powder or lumps
Assay (HPLC)≥98.0%
1-Pyrenecarboxaldehyde is a important intermediate used in agrochemical, pharmaceutical and dyestuff field. It is used as a highly polarity-sensitive fluorophore for SNP typing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Pyrenecarboxaldehyde is a important intermediate used in agrochemical, pharmaceutical and dyestuff field. It is used as a highly polarity-sensitive fluorophore for SNP typing.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong bases and strong oxidizing agents.
RUO – Research Use Only

General References:

  1. P. Lianos.; G. Cremel. ENVIRONMENTAL EFFECTS ON THE ELECTRONIC SPECTRAL PROPERTIES OF 1-PYRENECARBOXALDEHYDE AND THEIR APPLICATION IN PROBING BIOLOGICAL STRUCTURES. Photochemistry and Photobiology. 1980, 31 (5), 429-434.
  2. Fernando Secco.; Marcella Venturin.; Tarita Biver.;Francisco Sánchez.; Rafael Prado-Gotor.; Elia Grueso. Solvent Effects on the Kinetics of the Interaction of 1-Pyrenecarboxaldehyde with Calf Thymus DNA. J. Phys. Chem. B. 2010, 114 (13), 4686-4691.