2-Methoxyethyl isothiocyanate, 98+%
2-Methoxyethyl isothiocyanate, 98+%
2-Methoxyethyl isothiocyanate, 98+%
Thermo Scientific Chemicals

2-Methoxyethyl isothiocyanate, 98+%

CAS: 38663-85-3 | C4H7NOS | 117.166 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L10839.06
also known as L10839-06
Price (USD)
76.65
Online Exclusive
85.50
Save 8.85 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
76.65
Online Exclusive
85.50
Save 8.85 (10%)
Each
Chemical Identifiers
CAS38663-85-3
IUPAC Name1-isothiocyanato-2-methoxyethane
Molecular FormulaC4H7NOS
InChI KeyHTSAVXAFEVUJQE-UHFFFAOYSA-N
SMILESCOCCN=C=S
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to yellow to red to brown
FormCrystals or powder or crystalline powder or clear liquid as melt
Refractive Index1.5095-1.5140 @ 20?C
Assay (GC)≥98.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only