Ethyl 3-isothiocyanatopropionate, 97%
Ethyl 3-isothiocyanatopropionate, 97%
Ethyl 3-isothiocyanatopropionate, 97%
Thermo Scientific Chemicals

Ethyl 3-isothiocyanatopropionate, 97%

CAS: 17126-62-4 | C6H9NO2S | 159.203 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L10911.03
also known as L10911-03
Price (USD)
72.30
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
72.30
Each
Chemical Identifiers
CAS17126-62-4
IUPAC Nameethyl 3-isothiocyanatopropanoate
Molecular FormulaC6H9NO2S
InChI KeyUPTRONYNXNYITM-UHFFFAOYSA-N
SMILESCCOC(=O)CCN=C=S
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
FormLiquid
Assay (GC)≥96.0%
Refractive Index1.4995-1.5055 @ 20°C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only