Thymolphthalexone
Thymolphthalexone
Thymolphthalexone
Thermo Scientific Chemicals

Thymolphthalexone

CAS: 1913-93-5 | C38H44N2O12 | 720.77 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L10962.03
also known as L10962-03
Price (USD)
33.65
Online Exclusive
37.10
Save 3.45 (9%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
33.65
Online Exclusive
37.10
Save 3.45 (9%)
Each
Chemical Identifiers
CAS1913-93-5
IUPAC Name2-[({3-[1-(3-{[(carboxylatomethyl)(carboxymethyl)azaniumyl]methyl}-4-hydroxy-2-methyl-5-(propan-2-yl)phenyl)-3-oxo-1,3-dihydro-2-benzofuran-1-yl]-6-hydroxy-2-methyl-5-(propan-2-yl)phenyl}methyl)(carboxymethyl)azaniumyl]acetate
Molecular FormulaC38H44N2O12
InChI KeyAGXCADFSYJNMEG-UHFFFAOYNA-N
SMILESCC(C)C1=CC(=C(C)C(C[NH+](CC(O)=O)CC([O-])=O)=C1O)C1(OC(=O)C2=CC=CC=C12)C1=CC(C(C)C)=C(O)C(C[NH+](CC(O)=O)CC([O-])=O)=C1C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale brown
Formpowder
CommentSuitable for use as a complexiometric indicator
Color ChangeBlue in alkaline solution
Elemental AnalysisHydrogen 5.80 - 7.10% (Theory 6.153)
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only