Diallyl sulfide, 98%
Diallyl sulfide, 98%
Diallyl sulfide, 98%
Thermo Scientific Chemicals

Diallyl sulfide, 98%

CAS: 592-88-1 | C6H10S | 114.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number L10971.14
also known as L10971-14
Price (USD)
29.65
Special offer
Online exclusive
Ends: 30-Jun-2026
34.70
Save 5.05 (15%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
29.65
Special offer
Online exclusive
Ends: 30-Jun-2026
34.70
Save 5.05 (15%)
Each
Chemical Identifiers
CAS592-88-1
IUPAC Name3-(prop-2-en-1-ylsulfanyl)prop-1-ene
Molecular FormulaC6H10S
InChI KeyUBJVUCKUDDKUJF-UHFFFAOYSA-N
SMILESC=CCSCC=C
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Appearance (Color)Clear colorless
Assay (GC)≥97.5%
Refractive Index1.4870-1.4920 @ 20?C
Diallyl sulfide is a modulator of drug metabolizing enzyme P450 system and inducer of the phase II detoxifying enzyme GST. Diallyl sulfide is also an inhibitor of CYP2E1 and believed to prevent chemically-induced carcinogenesis in many tissues. It is also used as a food additive. It is also used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diallyl sulfide is a modulator of drug metabolizing enzyme P450 system and inducer of the phase II detoxifying enzyme GST. Diallyl sulfide is also an inhibitor of CYP2E1 and believed to prevent chemically-induced carcinogenesis in many tissues. It is also used as a food additive. It is also used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in alcohol, chloroform, ether, and carbon tetrachloride. Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
RUO – Research Use Only

General References:

  1. MJ Wargovich. Diallyl sulfide, a flavor component of garlic (Allium sativum), inhibits dimethyihydrazine-induced colon cancer. Carcinogenesis. 1987, 8,(3), 487-489.
  2. MJ Wargovich.; C Woods.; VWS Eng, LC Stephens. Chemoprevention of N-nitrosomethylbenzylamine-induced esophageal cancer in rats by the naturally occurring thioether, diallyl sulfide. Cancer Research. 1988, 48,(1), 6872-6875.