Phloretin, 98%
Phloretin, 98%
Phloretin, 98%
Thermo Scientific Chemicals

Phloretin, 98%

CAS: 60-82-2 | C15H14O5 | 274.272 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mg
100 mg
Catalog number L10991.MB
also known as L10991-MB
Price (USD)
37.65
Online Exclusive
41.50
Save 3.85 (9%)
Each
Quantity:
25 mg
Request bulk or custom format
Price (USD)
37.65
Online Exclusive
41.50
Save 3.85 (9%)
Each
Chemical Identifiers
CAS60-82-2
IUPAC Name3-(4-hydroxyphenyl)-1-(2,4,6-trihydroxyphenyl)propan-1-one
Molecular FormulaC15H14O5
InChI KeyVGEREEWJJVICBM-UHFFFAOYSA-N
SMILESOC1=CC=C(CCC(=O)C2=C(O)C=C(O)C=C2O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Cream or pale pink
FormCrystals or powder or crystalline powder
Assay (HPLC)≥97.5%
Topical antioxidant

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only