Methyltriphenylsilane, 97%
Methyltriphenylsilane, 97%
Methyltriphenylsilane, 97%
Thermo Scientific Chemicals

Methyltriphenylsilane, 97%

CAS: 791-29-7 | C19H18Si | 274.438 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number L10995.03
also known as L10995-03
Price (USD)
30.65
Online Exclusive
33.80
Save 3.15 (9%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
30.65
Online Exclusive
33.80
Save 3.15 (9%)
Each
Chemical Identifiers
CAS791-29-7
IUPAC Namemethyltriphenylsilane
Molecular FormulaC19H18Si
InChI KeyGIGVICQLYWGMGW-UHFFFAOYSA-N
SMILESC[Si](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream to cream to yellow
Melting Point (clear melt)61.0-70.0?C
FormCrystals or powder or crystalline powder
Assay (GC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only