4-Nitrophenyl isothiocyanate, 97%
4-Nitrophenyl isothiocyanate, 97%
4-Nitrophenyl isothiocyanate, 97%
Thermo Scientific Chemicals

4-Nitrophenyl isothiocyanate, 97%

CAS: 2131-61-5 | C7H4N2O2S | 180.181 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number L11042.30
also known as L11042-30
Price (USD)
495.65
Online Exclusive
551.00
Save 55.35 (10%)
Each
Quantity:
250 g
Request bulk or custom format
Price (USD)
495.65
Online Exclusive
551.00
Save 55.35 (10%)
Each
Chemical Identifiers
CAS2131-61-5
IUPAC Name1-isothiocyanato-4-nitrobenzene
Molecular FormulaC7H4N2O2S
InChI KeyNXHSSIGRWJENBH-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC=C(C=C1)N=C=S
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Cream to yellow
Identification (FTIR)Conforms
Melting Point (clear melt)106-112?C
FormPowder or lumpy solid
Assay (GC)≥96.0%
4-Nitrophenyl isothiocyanate is used as specializing in synthesizing of organic fine chemicals and pharmaceutical intermediates for use in development and production

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Nitrophenyl isothiocyanate is used as specializing in synthesizing of organic fine chemicals and pharmaceutical intermediates for use in development and production

Solubility
It hydrolyzes with water. Soluble in toluene.

Notes
Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
RUO – Research Use Only

General References:

  1. M Péter.; A Gyeresi.; F Fülöp. Liquid chromatographic enantioseparation of β-blocking agents with (1R, 2R)-1, 3-diacetoxy-1-(4-nitrophenyl)-2-propyl isothiocyanate as chiral derivatizing agent. Journal of Chromatography A. 2001, 910,(2), 247-253.
  2. YJ Kim.; H Kwak.; SJ Lee.; JS Lee.; HJ Kwon.; SH Nam. Urea/thiourea-based colorimetric chemosensors for the biologically important ions: efficient and simple sensors. Tetrahedron. 2006, 62,(41), 9635-9640.