4-Methylbenzotrifluoride, 98%
4-Methylbenzotrifluoride, 98%
4-Methylbenzotrifluoride, 98%
Thermo Scientific Chemicals

4-Methylbenzotrifluoride, 98%

CAS: 6140-17-6 | C8H7F3 | 160.139 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L11043.03
also known as L11043-03
Price (USD)
41.65
Online exclusive
46.30
Save 4.65
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
41.65
Online exclusive
46.30
Save 4.65
Each
Chemical Identifiers
CAS6140-17-6
IUPAC Name1-methyl-4-(trifluoromethyl)benzene
Molecular FormulaC8H7F3
InChI KeyLRLRAYMYEXQKID-UHFFFAOYSA-N
SMILESCC1=CC=C(C=C1)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear, colourless
Formliquid
Refractive Index1.4240-1.4280
Assay (GC)>97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only