Methyl 2-isothiocyanatoacetate, 98%
Methyl 2-isothiocyanatoacetate, 98%
Methyl 2-isothiocyanatoacetate, 98%
Methyl 2-isothiocyanatoacetate, 98%
Thermo Scientific Chemicals

Methyl 2-isothiocyanatoacetate, 98%

CAS: 21055-37-8 | C4H5NO2S | 131.149 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L11046.06
also known as L11046-06
Price (USD)
110.65
Special offer
Online exclusive
Ends: 30-Jun-2026
130.00
Save 19.35 (15%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
110.65
Special offer
Online exclusive
Ends: 30-Jun-2026
130.00
Save 19.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS21055-37-8
IUPAC Namemethyl 2-isothiocyanatoacetate
Molecular FormulaC4H5NO2S
InChI KeyGOWGDPFDGIPFIK-UHFFFAOYSA-N
SMILESCOC(=O)CN=C=S
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to orange to brown
FormLiquid
Assay (GC)≥96.0%
Refractive Index1.5170-1.5210 @ 20°C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only