2,4-Dichlorophenyl isocyanate, 96%
2,4-Dichlorophenyl isocyanate, 96%
2,4-Dichlorophenyl isocyanate, 96%
Thermo Scientific Chemicals

2,4-Dichlorophenyl isocyanate, 96%

CAS: 2612-57-9 | C7H3Cl2NO | 188.007 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number L11089.03
also known as L11089-03
Price (USD)
18.50
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
18.50
Each
Chemical Identifiers
CAS2612-57-9
IUPAC Name2,4-dichloro-1-isocyanatobenzene
Molecular FormulaC7H3Cl2NO
InChI KeyOLBJNSPBWLCTOT-UHFFFAOYSA-N
SMILESClC1=CC=C(N=C=O)C(Cl)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder or fused solid
Identification (FTIR)Conforms
Melting Point (clear melt)55.0-64.0?C
Assay (GC)≥95.0%
Appearance (Color)White to pale cream to pale brown
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only