3-Chloro-4-fluorothiobenzamide, 97%
3-Chloro-4-fluorothiobenzamide, 97%
3-Chloro-4-fluorothiobenzamide, 97%
Thermo Scientific Chemicals

3-Chloro-4-fluorothiobenzamide, 97%

CAS: 130560-97-3 | C7H5ClFNS | 189.632 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number L11132.MD
also known as L11132-MD
Price (USD)
40.65
Special offer
Online exclusive
Ends: 30-Jun-2026
47.50
Save 6.85 (14%)
Each
Quantity:
250 mg
Request bulk or custom format
Price (USD)
40.65
Special offer
Online exclusive
Ends: 30-Jun-2026
47.50
Save 6.85 (14%)
Each
Chemical Identifiers
CAS130560-97-3
IUPAC Name3-chloro-4-fluorobenzene-1-carbothioamide
Molecular FormulaC7H5ClFNS
InChI KeyRSGPCKCTKDCMRR-UHFFFAOYSA-N
SMILESNC(=S)C1=CC(Cl)=C(F)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)≥96.0%
Appearance (Color)Yellow
FormPowder or lumps
Melting Point (clear melt)126.0-132.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only