4-Methylbenzyl isothiocyanate, 96%
4-Methylbenzyl isothiocyanate, 96%
4-Methylbenzyl isothiocyanate, 96%
Thermo Scientific Chemicals

4-Methylbenzyl isothiocyanate, 96%

CAS: 3694-46-0 | C9H9NS | 163.238 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
10 g
Catalog number L11135.04
also known as L11135-04
Price (USD)
31.50
Each
Quantity:
2 g
Request bulk or custom format
Price (USD)
31.50
Each
Chemical Identifiers
CAS3694-46-0
IUPAC Name1-(isothiocyanatomethyl)-4-methylbenzene
Molecular FormulaC9H9NS
InChI KeyOAXIUBJXQISJEV-UHFFFAOYSA-N
SMILESCC1=CC=C(CN=C=S)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
CommentSpecification differs for U.S. and non-U.S. material where indicated
Proton NMRConforms to structure (U.S. specification)
Refractive Index1.5870-1.5940 @ 20°C (non-U.S. specification)
Assay (GC)≥94.0%
Appearance (Color)Clear yellow to orange to brown
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only