CommentSpecification differs for U.S. and non-U.S. material where indicated
Proton NMRConforms to structure (U.S. specification)
Refractive Index1.5870-1.5940 @ 20°C (non-U.S. specification)
Assay (GC)≥94.0%
Appearance (Color)Clear yellow to orange to brown
View more