(S)-(-)-1-Phenylethyl isocyanate, 98%
(S)-(-)-1-Phenylethyl isocyanate, 98%
(S)-(-)-1-Phenylethyl isocyanate, 98%
Thermo Scientific Chemicals

(S)-(-)-1-Phenylethyl isocyanate, 98%

CAS: 14649-03-7 | C9H9NO | 147.177 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L11141.06
also known as L11141-06
Price (USD)
187.65
Special offer
Online exclusive
Ends: 30-Jun-2026
221.00
Save 33.35 (15%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
187.65
Special offer
Online exclusive
Ends: 30-Jun-2026
221.00
Save 33.35 (15%)
Each
Chemical Identifiers
CAS14649-03-7
IUPAC Name(1-isocyanatoethyl)benzene
Molecular FormulaC9H9NO
InChI KeyJJSCUXAFAJEQGB-UHFFFAOYNA-N
SMILESCC(N=C=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to pale yellow
FormLiquid
Assay (GC)≥97.5%
Proton NMRConforms
(S)-(-)-1-Phenylethyl isocyanate act by inhibition of cell proliferation and induction of apoptosis. It helps to prevent lung cancer and esophageal cancer. It is a fine chemical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-(-)-1-Phenylethyl isocyanate act by inhibition of cell proliferation and induction of apoptosis. It helps to prevent lung cancer and esophageal cancer. It is a fine chemical intermediate.

Solubility
Hydrolyzes in water.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers. Containers which are opened must be carefully resealed and kept upright to prevent leakage. It is sensitive to moisture. Recommended storage temperature is 2 - 8°C.
RUO – Research Use Only
Yoshio Okamoto.; Yuriko Kaida.; Hiromi Hayashida.; Koichi Hatada. Tris(1-phenylethylcarbamate)s of Cellulose and Amylose as Useful Chiral Stationary Phases for Chromatographic Optical Resolution. Chemistry Letters. 1990, 19 (6), 909-912.