1,1,1-Trifluoro-2-propanol, 97%
1,1,1-Trifluoro-2-propanol, 97%
1,1,1-Trifluoro-2-propanol, 97%
Thermo Scientific Chemicals

1,1,1-Trifluoro-2-propanol, 97%

CAS: 374-01-6 | C3H5F3O | 114.067 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L11150.06
also known as L11150-06
Price (USD)
61.65
Online Exclusive
68.60
Save 6.95 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
61.65
Online Exclusive
68.60
Save 6.95 (10%)
Each
Chemical Identifiers
CAS374-01-6
IUPAC Name1,1,1-trifluoropropan-2-ol
Molecular FormulaC3H5F3O
InChI KeyGILIYJDBJZWGBG-UHFFFAOYNA-N
SMILESCC(O)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)≥96.0%
CommentSpecification differs for U.S. and non-U.S. material where indicated
Identification (FTIR)Conforms
View more
1,1,1-Trifluoro-2-propanol is used as a chemical building block.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,1,1-Trifluoro-2-propanol is used as a chemical building block.

Solubility
Fully miscible in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with bases and oxidizing agents.
RUO – Research Use Only

General References:

  1. Toshimasa Katagiri.; Hideki Ihara.; Mikihiro Takahashi.; Setsuo Kashino.; Keizo Furuhashi.; Kenji Uneyama. Intramolecular SN2 reaction at α-carbon of trifluoromethyl group: preparation of optically active 2-trifluoromethylaziridine. Tetrahedron: Asymmetry. 1977, 8 (17), 2933-2937.
  2. Juhani Murto.; Antti Kivinen.; Kari Edelmann.; Eero Hassinen. Fluoroalcohols—XXI. Infrared, matrix infrared and Raman spectra of 1,1,1-trifluoro-2-propanol and its three deuterated analogues. Spectrochimica Acta Part A: Molecular Spectroscopy. 1975, 31 (4), 479-493.