3-Butynyl p-toluenesulfonate, 96%
3-Butynyl p-toluenesulfonate, 96%
3-Butynyl p-toluenesulfonate, 96%
Thermo Scientific Chemicals

3-Butynyl p-toluenesulfonate, 96%

CAS: 23418-85-1 | C11H12O3S | 224.274 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L11166.06
also known as L11166-06
Price (USD)
97.65
Online Exclusive
108.00
Save 10.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
97.65
Online Exclusive
108.00
Save 10.35 (10%)
Each
Chemical Identifiers
CAS23418-85-1
IUPAC Namebut-3-yn-1-yl 4-methylbenzene-1-sulfonate
Molecular FormulaC11H12O3S
InChI KeySTOASOOVVADOKH-UHFFFAOYSA-N
SMILESCC1=CC=C(C=C1)S(=O)(=O)OCCC#C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow to orange to dark brown to brown
FormLiquid or viscous liquid
Assay (GC)≥95.0%
Refractive Index1.5215-1.5275 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only