6-Aminocoumarin hydrochloride, 97%
6-Aminocoumarin hydrochloride, 97%
6-Aminocoumarin hydrochloride, 97%
Thermo Scientific Chemicals

6-Aminocoumarin hydrochloride, 97%

CAS: 63989-79-7 | C9H8ClNO2 | 197.62 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L11168.03
also known as L11168-03
Price (USD)
63.65
Special offer
Online exclusive
Ends: 28-Dec-2026
74.80
Save 11.15
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
63.65
Special offer
Online exclusive
Ends: 28-Dec-2026
74.80
Save 11.15
Each
Chemical Identifiers
CAS63989-79-7
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to dark yellow to brown to pale gray
FormCrystals or powder or crystalline powder or fused solid or granules
Assay (HPLC)≥96.0%
Assay (Titration ex Chloride)≥96.0 to ≤104.0%
6-Aminocoumarin hydrochloride is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Aminocoumarin hydrochloride is used as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with bases and oxidizing agents.
RUO – Research Use Only

General References:

  1. Robert C. Elderfield.; J. Roy. Synthesis of Potential Anticancer Agents. XVIII. Nitrogen Mustards from 6-Substituted Coumarins. J. Med. Chem. 1967, 10 (5), 918-921.
  2. Suman Roy.; Tapan Kumar Mondal.; Partha Mitra.; Elena Lopez Torres.; Chittaranjan Sinha. Synthesis, structure, spectroscopic properties, electrochemistry, and DFT correlative studies of N-[(2-pyridyl)methyliden]-6-coumarin complexes of Cu(I) and Ag(I). Polyhedron. 2011, 30 (6), 913-922.