5-Bromoindoxyl diacetate, 98+%, Thermo Scientific Chemicals
5-Bromoindoxyl diacetate, 98+%, Thermo Scientific Chemicals
5-Bromoindoxyl diacetate, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5-Bromoindoxyl diacetate, 98+%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
L11189.03
also known as L11189-03
1 g
Catalog number L11189.03
also known as L11189-03
Price (USD)
57.65
Special offer
Online exclusive
Ends: 28-Dec-2026
67.50
Save 9.85
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
57.65
Special offer
Online exclusive
Ends: 28-Dec-2026
67.50
Save 9.85
Each
Chemical Identifiers
CAS33588-54-4
IUPAC Name1-acetyl-5-bromo-1H-indol-3-yl acetate
Molecular FormulaC12H10BrNO3
InChI KeyXJRIDJAGAYGJCK-UHFFFAOYSA-N
SMILESCC(=O)OC1=CN(C(C)=O)C2=CC=C(Br)C=C12
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Appearance (Color)White to cream to yellow
Assay (GC)≥98.0%
Melting Point (clear melt)121.5-127.5?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only