1,2-Benzenedithiol, 97%
1,2-Benzenedithiol, 97%
1,2-Benzenedithiol, 97%
Thermo Scientific Chemicals

1,2-Benzenedithiol, 97%

CAS: 17534-15-5 | C6H6S2 | 142.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number L11253.06
Price (USD)
494.65
Online Exclusive
549.00
Save 54.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
494.65
Online Exclusive
549.00
Save 54.35 (10%)
Each
Chemical Identifiers
CAS17534-15-5
IUPAC Namebenzene-1,2-dithiol
Molecular FormulaC6H6S2
InChI KeyJRNVQLOKVMWBFR-UHFFFAOYSA-N
SMILESSC1=C(S)C=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.6555-1.6605 as melt
Appearance (Color)Clear colorless to yellow
Assay (GC)>96.0%
FormLiquid as melt
1,2-Benzenedithiol is used in preparation of schiff base, which is employed to make new series of copper(II) and zinc(II) complexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2-Benzenedithiol is used in preparation of schiff base, which is employed to make new series of copper(II) and zinc(II) complexes.

Solubility
Soluble in water, benzene, ethanol, diethyl ether, ethyl acetate.

Notes
Incompatible with bases and strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Giguere, J.; Morin, J. Superextended Tetrathiafulvalene: Synthesis, Optoelectronic Properties, Fullerenes Complexation, and Photooxidation Study. J. Org. Chem. 2015, 80 (13), 6767-6775.
  2. Nouchi, R.; Tanimoto, T. Substituent-Controlled Reversible Switching of Charge Injection Barrier Heights at Metal/Organic Semiconductor Contacts Modified with Disordered Molecular Monolayers. ACS Nano. 2015, 9 (7), 7429-7439.