4-n-Butoxybenzenesulfonyl chloride, 97%
4-n-Butoxybenzenesulfonyl chloride, 97%
4-n-Butoxybenzenesulfonyl chloride, 97%
Thermo Scientific Chemicals

4-n-Butoxybenzenesulfonyl chloride, 97%

CAS: 1138-56-3 | C10H13ClO3S | 248.721 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L11258.06
also known as L11258-06
Price (USD)
205.65
Special offer
Online exclusive
Ends: 30-Jun-2026
242.00
Save 36.35 (15%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
205.65
Special offer
Online exclusive
Ends: 30-Jun-2026
242.00
Save 36.35 (15%)
Each
Specifications
Beilstein1109264
CAS1138-56-3
Chemical Name or Material4-n-Butoxybenzenesulfonyl chloride
DOT InformationTransport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECS Number000-000-0
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only