6-Dodecyne, 98%
6-Dodecyne, 98%
6-Dodecyne, 98%
Thermo Scientific Chemicals

6-Dodecyne, 98%

CAS: 6975-99-1 | C12H22 | 166.31 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L11262.06
also known as L11262-06
Price (USD)
77.60
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
77.60
Each
Chemical Identifiers
CAS6975-99-1
IUPAC Namedodec-6-yne
Molecular FormulaC12H22
InChI KeyIHPFQAOOSAGSPN-UHFFFAOYSA-N
SMILESCCCCCC#CCCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥97.5% (non-U.S. Specification)
Assay from Suppliers CofA≥97.5% (GC: U.S. specification)
CommentSpecification differs for U.S. and non-U.S. material where indicated
Refractive Index1.4375-1.4420 @ 20°C (non-U.S. specification)
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only