2-Amino-4-methoxybenzothiazole, 97%
2-Amino-4-methoxybenzothiazole, 97%
2-Amino-4-methoxybenzothiazole, 97%
Thermo Scientific Chemicals

2-Amino-4-methoxybenzothiazole, 97%

CAS: 5464-79-9 | C8H8N2OS | 180.225 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number L11358.03
also known as L11358-03
Price (USD)
44.65
Special offer
Online exclusive
Ends: 30-Jun-2026
52.70
Save 8.05 (15%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
44.65
Special offer
Online exclusive
Ends: 30-Jun-2026
52.70
Save 8.05 (15%)
Each
Chemical Identifiers
CAS5464-79-9
IUPAC Name4-methoxy-1,3-benzothiazol-2-amine
Molecular FormulaC8H8N2OS
InChI KeyYEBCRAVYUWNFQT-UHFFFAOYSA-N
SMILESCOC1=C2N=C(N)SC2=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to pale yellow
Assay (GC)≥96.0%
FormCrystals or powder or crystalline powder
Melting Point (clear melt)150.0-159.0?C
Assay (Non-aqueous acid-base Titration)≥96.0 to ≤104.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only