3-Bromophenylurea, 97%
3-Bromophenylurea, 97%
3-Bromophenylurea, 97%
Thermo Scientific Chemicals

3-Bromophenylurea, 97%

CAS: 2989-98-2 | C7H7BrN2O | 215.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L11370.06
also known as L11370-06
Price (USD)
92.65
Online Exclusive
103.00
Save 10.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
92.65
Online Exclusive
103.00
Save 10.35 (10%)
Each
Specifications
Beilstein2718463
CAS2989-98-2
Chemical Name or Material3-Bromophenylurea
EINECS Number000-000-0
Formula Weight215.06
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only