Ethyl 4,4,4-trifluorobutyrate, 98%
Ethyl 4,4,4-trifluorobutyrate, 98%
Ethyl 4,4,4-trifluorobutyrate, 98%
Thermo Scientific Chemicals

Ethyl 4,4,4-trifluorobutyrate, 98%

CAS: 371-26-6 | C6H9F3O2 | 170.131 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L11399.03
also known as L11399-03
Price (USD)
23.70
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
23.70
Each
Chemical Identifiers
CAS371-26-6
IUPAC Nameethyl 4,4,4-trifluorobutanoate
Molecular FormulaC6H9F3O2
InChI KeyPSRZMXNNQTWAGB-UHFFFAOYSA-N
SMILESCCOC(=O)CCC(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colourless
FormLiquid
Refractive Index1.3200-1.3540 @20?C
Assay (GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only