4-Chlorocinnamamide, 97%
4-Chlorocinnamamide, 97%
4-Chlorocinnamamide, 97%
Thermo Scientific Chemicals

4-Chlorocinnamamide, 97%

CAS: 18166-64-8 | C9H8ClNO | 181.62 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L11406.03
also known as L11406-03
Price (USD)
61.65
Special offer
Online exclusive
Ends: 28-Dec-2026
72.70
Save 11.05
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
61.65
Special offer
Online exclusive
Ends: 28-Dec-2026
72.70
Save 11.05
Each
Chemical Identifiers
CAS18166-64-8
IUPAC Name(2E)-3-(4-chlorophenyl)prop-2-enamide
Molecular FormulaC9H8ClNO
InChI KeyPWXPFYVNYKVJBW-ZZXKWVIFSA-N
SMILESNC(=O)C=CC1=CC=C(Cl)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale brown
FormCrystals or powder or crystalline powder
Melting Point (clear melt)210.0-219.0?C
Assay (HPLC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only