Methyl 4,4,4-trifluorocrotonate, 97%
Methyl 4,4,4-trifluorocrotonate, 97%
Methyl 4,4,4-trifluorocrotonate, 97%
Thermo Scientific Chemicals

Methyl 4,4,4-trifluorocrotonate, 97%

CAS: 85694-31-1 | C5H5F3O2 | 154.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L11466.06
also known as L11466-06
Price (USD)
183.65
Online Exclusive
204.00
Save 20.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
183.65
Online Exclusive
204.00
Save 20.35 (10%)
Each
Chemical Identifiers
CAS85694-31-1
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.3510-1.3560 @ 20?C
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)>96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only