3,5-Dihydroxybenzhydrazide, 98%
3,5-Dihydroxybenzhydrazide, 98%
3,5-Dihydroxybenzhydrazide, 98%
Thermo Scientific Chemicals

3,5-Dihydroxybenzhydrazide, 98%

CAS: 7732-32-3 | C7H8N2O3 | 168.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L11471.14
also known as L11471-14
Price (USD)
218.65
Special offer
Online exclusive
Ends: 30-Jun-2026
257.00
Save 38.35 (15%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
218.65
Special offer
Online exclusive
Ends: 30-Jun-2026
257.00
Save 38.35 (15%)
Each
Chemical Identifiers
CAS7732-32-3
IUPAC Name3,4-dihydroxybenzohydrazide
Molecular FormulaC7H8N2O3
InChI KeyWGXWEXNJRZMIPT-UHFFFAOYSA-N
SMILESNNC(=O)C1=CC=C(O)C(O)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Appearance (Color)White to cream
Assay (HPLC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only