3-Hydroxyphenylacetylene, 97%
3-Hydroxyphenylacetylene, 97%
3-Hydroxyphenylacetylene, 97%
3-Hydroxyphenylacetylene, 97%
Thermo Scientific Chemicals

3-Hydroxyphenylacetylene, 97%

CAS: 10401-11-3 | C8H6O | 118.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L11523.03
also known as L11523-03
Price (USD)
86.65
Online Exclusive
96.60
Save 9.95 (10%)
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
86.65
Online Exclusive
96.60
Save 9.95 (10%)
Each
Add to cart
Chemical Identifiers
CAS10401-11-3
IUPAC Name3-ethynylphenol
Molecular FormulaC8H6O
InChI KeyAODMJIOEGCBUQL-UHFFFAOYSA-N
SMILESOC1=CC=CC(=C1)C#C
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)>96.0%
Appearance (Color)Clear colorless to yellow or brown
Refractive Index1.5845-1.5895 @ 20?C
FormLiquid
3-Hydroxyphenylacetylene is used as a fluorogenic and chromogenic probe for bacterial enzymes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Hydroxyphenylacetylene is used as a fluorogenic and chromogenic probe for bacterial enzymes.

Solubility
Miscible with dimethylformamide.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Bassetti, M.; Ciceri, S.; Lancia, F.; Pasquini, C. Hydration of aromatic terminal alkynes catalyzed by iron(III) sulfate hydrate under chlorine-free conditions. Tetrahedron Lett. 2014, 55 (9), 1608-1612.
  2. Jiang, Q.; Zhao, A.; Xu, B.; Jia, J.; Liu, X.; Guo, C. PIFA-Mediated Esterification Reaction of Alkynes with Alcohols via Oxidative Cleavage of Carbon Triple Bonds. J. Org. Chem. 2014, 79 (6), 2709-2715.