1-Benzhydrylpiperazine, 97%
1-Benzhydrylpiperazine, 97%
1-Benzhydrylpiperazine, 97%
1-Benzhydrylpiperazine, 97%
Thermo Scientific Chemicals

1-Benzhydrylpiperazine, 97%

CAS: 841-77-0 | C17H20N2 | 252.361 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number L11530.22
also known as L11530-22
Price (USD)
80.65
Online Exclusive
89.90
Save 9.25 (10%)
Each
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)
80.65
Online Exclusive
89.90
Save 9.25 (10%)
Each
Add to cart
Chemical Identifiers
CAS841-77-0
IUPAC Name1-(diphenylmethyl)piperazin-1-ium chloride
Molecular FormulaC17H21ClN2
InChI KeyDLCXUFKAJDKSDD-UHFFFAOYSA-N
SMILES[Cl-].C1C[NH+](CCN1)C(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Assay (GC)≥96.0%
Appearance (Color)White to cream to pale yellow
Melting Point (clear melt)87.0-96.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only