2-Acetoxyisobutyryl bromide, 96%
2-Acetoxyisobutyryl bromide, 96%
2-Acetoxyisobutyryl bromide, 96%
Thermo Scientific Chemicals

2-Acetoxyisobutyryl bromide, 96%

CAS: 40635-67-4 | C6H9BrO3 | 209.039 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number L11592.22
also known as L11592-22
Price (USD)
214.00
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
214.00
Each
Chemical Identifiers
CAS40635-67-4
IUPAC Name1-bromo-2-methyl-1-oxopropan-2-yl acetate
Molecular FormulaC6H9BrO3
InChI KeyOOKAXSHFTDPZHP-UHFFFAOYSA-N
SMILESCC(=O)OC(C)(C)C(Br)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow to brown or pale purple
FormLiquid
Assay (Titration ex Bromide)≥95.0 to ≤105.0%
Refractive Index1.4530-1.4595 @ 20°C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only