3-(Trifluoromethyl)phenyl isothiocyanate, 98%
3-(Trifluoromethyl)phenyl isothiocyanate, 98%
3-(Trifluoromethyl)phenyl isothiocyanate, 98%
3-(Trifluoromethyl)phenyl isothiocyanate, 98%
Thermo Scientific Chemicals

3-(Trifluoromethyl)phenyl isothiocyanate, 98%

CAS: 1840-19-3 | C8H4F3NS | 203.182 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number L11646.14
also known as L11646-14
Price (USD)
166.65
Online Exclusive
185.00
Save 18.35 (10%)
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
166.65
Online Exclusive
185.00
Save 18.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS1840-19-3
IUPAC Name1-isothiocyanato-3-(trifluoromethyl)benzene
Molecular FormulaC8H4F3NS
InChI KeyGFEPANUKFYVALF-UHFFFAOYSA-N
SMILESFC(F)(F)C1=CC=CC(=C1)N=C=S
View more
SpecificationsSpecification SheetSpecification Sheet
CommentSpecification differs for U.S. and non-U.S. material where indicated
Identification (FTIR)Conforms (non-U.S. specification)
Refractive Index1.5530-1.5570 @ 20°C (non-U.S. specification)
Appearance (Color)Clear colorless to yellow
FormLiquid
View more
Used as pharamaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as pharamaceutical intermediate.

Solubility
Hydrolyzes in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to moisture.
RUO – Research Use Only

General References:

  1. Ha-Young Kim.; Se Hun Kwak.; Gee-Hyung Lee.; Young-Dae Gong. Copper-catalyzed synthesis of 3-substituted-5-amino-1,2,4-thiadiazoles via intramolecular N-S bond formation. Tetrahedron. 2014, 70 (45), 8737-8743.
  2. Sara Van Poecke.; Hélène Munier-Lehmann.; Olivier Helynck.; Matheus Froeyend.; Serge Van Calenbergh. Synthesis and inhibitory activity of thymidine analogues targeting Mycobacterium tuberculosis thymidine monophosphate kinase. Bioorganic & Medicinal Chemistry. 2011, 19 (24), 7603-7611.