2-Methoxyphenyl isothiocyanate, 98%
Thermo Scientific Chemicals

2-Methoxyphenyl isothiocyanate, 98%

CAS: 3288-04-8 | C8H7NOS | 165.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L11799.06
also known as L11799-06
Price (USD)
55.65
Online Exclusive
61.60
Save 5.95 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
55.65
Online Exclusive
61.60
Save 5.95 (10%)
Each
Chemical Identifiers
CAS3288-04-8
IUPAC Name1-isothiocyanato-2-methoxybenzene
Molecular FormulaC8H7NOS
InChI KeyQKAOOWJWWKWWOZ-UHFFFAOYSA-N
SMILESCOC1=CC=CC=C1N=C=S
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
FormLiquid
Assay (GC)≥97.5%
Identification (FTIR)Conforms
Refractive Index1.6420-1.6470 @ 20°C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Solubility
Reacts with water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents, amines ansd alcohol. It is sensitive to moisture.
RUO – Research Use Only

General References:

  1. Jayashree Yenagi.; Anita R. Nandurkar.; J. Tonannavar. 2-Methoxyphenyl isocyanate and 2-Methoxyphenyl isothiocyanate: Conformers, vibration structure and multiplet Fermi resonance. Spectrochimica Acta Part A: Molecular and Biomolecular Spectroscopy. 2012, 91 261-268.
  2. L. Valettea.; S. Poulain.; X. Fernandez.; L. Lizzani-Cuvelier. Efficient and solvent-free microwave-accelerated synthesis of isothiocyanates using Lawesson’s reagent. Journal of Sulfur Chemistry. 2005, 26 (2), 155-161.