(+/-)-1-(4-Bromophenyl)ethylamine, 96%
(+/-)-1-(4-Bromophenyl)ethylamine, 96%
(+/-)-1-(4-Bromophenyl)ethylamine, 96%
Thermo Scientific Chemicals

(+/-)-1-(4-Bromophenyl)ethylamine, 96%

CAS: 24358-62-1 | C8H10BrN | 200.079 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L11848.06
also known as L11848-06
Price (USD)
52.65
Special offer
Online exclusive
Ends: 30-Jun-2026
61.30
Save 8.65 (14%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
52.65
Special offer
Online exclusive
Ends: 30-Jun-2026
61.30
Save 8.65 (14%)
Each
Chemical Identifiers
CAS24358-62-1
IUPAC Name(1R)-1-(4-bromophenyl)ethan-1-aminium
Molecular FormulaC8H11BrN
InChI KeySOZMSEPDYJGBEK-ZCFIWIBFSA-O
SMILESC[C@@H]([NH3+])C1=CC=C(Br)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.5640-1.5700 @ 20?C
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥95.0%
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only