1-(4-Fluorophenyl)-2-methyl-2-propylamine, 96%
1-(4-Fluorophenyl)-2-methyl-2-propylamine, 96%
1-(4-Fluorophenyl)-2-methyl-2-propylamine, 96%
Thermo Scientific Chemicals

1-(4-Fluorophenyl)-2-methyl-2-propylamine, 96%

CAS: 1200-27-7 | C10H14FN | 167.227 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L11858.06
also known as L11858-06
Price (USD)
314.65
Special offer
Online exclusive
Ends: 30-Jun-2026
370.00
Save 55.35 (15%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
314.65
Special offer
Online exclusive
Ends: 30-Jun-2026
370.00
Save 55.35 (15%)
Each
Chemical Identifiers
CAS1200-27-7
IUPAC Name1-(4-fluorophenyl)-2-methylpropan-2-amine
Molecular FormulaC10H14FN
InChI KeyJITFIYFVPMQJOK-UHFFFAOYSA-N
SMILESCC(C)(N)CC1=CC=C(F)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofA≥95.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only