Pentafluorophenyl isothiocyanate, 96%
Pentafluorophenyl isothiocyanate, 96%
Pentafluorophenyl isothiocyanate, 96%
Pentafluorophenyl isothiocyanate, 96%
Thermo Scientific Chemicals

Pentafluorophenyl isothiocyanate, 96%

CAS: 35923-79-6 | C7F5NS | 225.136 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L11874.03
also known as L11874-03
Price (USD)
85.65
Online Exclusive
94.80
Save 9.15 (10%)
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
85.65
Online Exclusive
94.80
Save 9.15 (10%)
Each
Add to cart
Chemical Identifiers
CAS35923-79-6
IUPAC Name1,2,3,4,5-pentafluoro-6-isothiocyanatobenzene
Molecular FormulaC7F5NS
InChI KeyNGNKMRBGZPDABE-UHFFFAOYSA-N
SMILESFC1=C(F)C(F)=C(N=C=S)C(F)=C1F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear, colorless to yellow or pink to brown
Refractive Index1.5350-1.5410 @ 20°C
FormLiquid
Assay (GC)≥95.0%
Pentafluorophenyl isothiocyanate is used in the preparation of 5-isopropyl-3-pentafluorophenyl-2-thiohydantoin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pentafluorophenyl isothiocyanate is used in the preparation of 5-isopropyl-3-pentafluorophenyl-2-thiohydantoin.

Notes
Moisture sensitive. Store in a cool place. Incompatible with strong oxidizing agents, strong acids and bases.
RUO – Research Use Only

General References:

  1. Kotova, N.; Frostne, C.; Abramsson-Zetterberg, L.; Tareke, E.; Bergman, R.; Haghdoost, S.; Paulsson, B.; Törnqvist, M.; Segerbäck, D.; Jenssen, D.; Grawé, J. Differences in micronucleus frequency and acrylamide adduct levels with hemoglobin between vegetarians and non-vegetarians. Eur. J. Nutr. 2014, 54 (7), 1181-1190.
  2. Scherer, G.; Newland, K.; Papadopoulou, E.; Minet, E. A correlation study applied to biomarkers of internal and effective dose for acrylonitrile and 4-aminobiphenyl in smokers. Biomarkers 2014, 19 (4), 291-301.